Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golim4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291069.1 | NP_001277998.1 | 5 | 1151 | ||
NM_175193.6 | NP_780402.1 | 5 | 1151 | |||
GenBank mRNA | AK033730.1 | - | 5 | 1138 | ||
AK048434.1 | - | 6 | 1062 | |||
AK078928.1 | - | 5 | 1151 | |||
AK149286.1 | - | 5 | 1111 | |||
BC086642.1 | - | 5 | 983 | 6836803 | ||
BC089354.1 | - | 5 | 1033 | 6837211 |