- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Psapl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175249.3 | NP_780458.2 | 1 | 2137 | ||
GenBank mRNA | AK009408.1 | - | 1 | 2125 | ||
AK028455.1 | - | 1 | 2102 | |||
AK078699.1 | - | 1 | 2105 | |||
AV088427.2 | - | 1 | 648 |