Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175319.4 | NP_780528.1 | 4 | 670 | ||
GenBank mRNA | AK049131.1 | - | 4 | 671 | ||
BC094908.1 | - | 4 | 566 | 30021730 | ||
BC132375.1 | - | 4 | 657 | 40130652 | ||
BC138307.1 | - | 4 | 656 | 8861327 |