Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bola3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175277.4 | NP_780486.1 | 3, 4 | 292 | ||
GenBank mRNA | AK019037.1 | - | 2, 3 | 127 | ||
AK028102.1 | - | 3, 4 | 293 | |||
AK131950.1 | - | 301 | ||||
AK160488.1 | - | 3, 4 | 296 | |||
AK161878.1 | - | 64 | ||||
AK218122.1 | - | 1, 2 | 109 | |||
BC049748.1 | - | 3, 4 | 312 | |||
BC116947.1 | - | 4, 5 | 330 | 40126266 | ||
BC116949.1 | - | 4, 5 | 330 | 40126268 |