Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175333.3 | NP_780542.1 | 6, 7 | 808 | ||
GenBank mRNA | AK077111.1 | - | 6, 7 | 809 | ||
BC115588.1 | - | 7, 8 | 907 | 40102382 | ||
BC115589.1 | - | 6, 7 | 801 | 40102384 | ||
FM165286.1 | - | 6, 7 | 750 |