Gene Symbol: Fzd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175284.3 | NP_780493.1 | 1 | 1786 | ||
GenBank mRNA | AF310970.1 | - | 1 | 1960 | ||
AK052950.1 | - | 1 | 2049 | |||
AY509002.1 | - | 1 | 1786 | |||
BC117759.2 | - | 1 | 1749 | 40090826 |
Set Membership: |
![]() |