Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmnat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175460.3 | NP_780669.1 | 2 | 111 | ||
GenBank mRNA | AK083532.1 | - | 2 | 215 | ||
AK084041.1 | - | 2 | 221 | |||
AK129155.1 | - | 2 | 197 | |||
BC089007.1 | - | 2 | 111 | 30532596 |