Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175484.3 | NP_780693.2 | 4, 5 | 868 | ||
GenBank mRNA | AK047527.1 | - | 4, 5 | 810 | ||
AK087414.1 | - | 4, 5 | 869 | |||
BC062649.1 | - | 4, 5 | 600 | 6416110 |