Gene Symbol: Lhfp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175386.3 | NP_780595.1 | 4 | 1268 | ||
GenBank mRNA | AK034532.1 | - | 4 | 1106 | ||
AK159126.1 | - | 4 | 1257 | |||
AK159549.1 | - | 4 | 1267 | |||
AK171791.1 | - | 4 | 729 | |||
AK172390.1 | - | 4 | 729 | |||
AK172532.1 | - | 4 | 729 | |||
BC052079.1 | - | 4 | 1268 | 6402782 | ||
BC095934.1 | - | 4 | 1112 | 3154659 |