Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175439.3 | NP_780648.1 | 1 | 1220 | ||
GenBank mRNA | AK042888.1 | - | 1 | 1161 | ||
AK050196.1 | - | 1 | 1177 | |||
AK144628.1 | - | 1 | 1161 | |||
AK172240.1 | - | 1 | 582 | |||
BC099669.1 | - | 1 | 1194 | 3153138 | ||
BC132343.1 | - | 1 | 1273 | 40130620 | ||
BC137739.1 | - | 1 | 1273 | 8860758 |