Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175506.3 | NP_780715.1 | 2 | 402 | ||
GenBank mRNA | AK053213.1 | - | 2 | 774 | ||
AK165144.1 | - | 1 | 389 | |||
AY135183.1 | - | 2 | 402 |