Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175522.3 | NP_780731.1 | 2 | 1320 | ||
GenBank mRNA | AK044511.1 | - | 2 | 1321 | ||
BC059029.1 | - | 1 | 1052 | 6811341 |