Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nim1k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175538.3 | NP_780747.1 | 3 | 1195 | ||
GenBank mRNA | AK044462.1 | - | 2 | 900 | ||
AK047794.1 | - | 3 | 1185 | |||
AK048694.1 | - | 3 | 1196 | |||
AK053734.1 | - | 3 | 1202 | |||
BC051189.1 | - | 2 | 862 | 6491737 | ||
BC106122.1 | - | 2 | 889 | 4505882 | ||
BC117955.1 | - | 3 | 1121 | 40098149 | ||
BC117956.1 | - | 3 | 1119 | 40098152 |