Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nlrp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175532.3 | NP_780741.1 | 3 | 1647 | ||
GenBank mRNA | AK028575.1 | - | 3 | 1670 | ||
AK028802.1 | - | 3 | 1649 | |||
AK032446.1 | - | 3 | 1643 | |||
AY355344.1 | - | 2 | 1616 | |||
AY489193.1 | - | 2 | 1600 | |||
BC113194.1 | - | 3 | 1643 | 40062457 | ||
BC113801.1 | - | 3 | 1643 | 40062458 |