Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmrta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175647.3 | NP_783578.1 | 2 | 3079 | ||
GenBank mRNA | AF542047.1 | - | 2 | 3079 | ||
BC137613.1 | - | 2 | 3013 | 8860632 |