Search Thermo Fisher Scientific
Gene Symbol: ICK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014920.3 | NP_055735.1 | 14 | 3658 | ||
NM_016513.4 | NP_057597.2 | 15 | 3769 | |||
XM_011514419.1 | XP_011512721.1 | 14 | 3679 | |||
XM_011514420.2 | XP_011512722.1 | 15 | 3821 | |||
XM_011514421.1 | XP_011512723.1 | 15 | 3760 | |||
XM_017010485.1 | XP_016865974.1 | 15 | 4053 | |||
XM_017010486.1 | XP_016865975.1 | 16 | 3932 | |||
XM_017010487.1 | XP_016865976.1 | 14 | 3598 | |||
XM_017010488.1 | XP_016865977.1 | 15 | 4459 | |||
XM_017010489.1 | XP_016865978.1 | 16 | 3974 | |||
XM_017010490.1 | XP_016865979.1 | 15 | 3844 | |||
XM_017010491.1 | XP_016865980.1 | 15 | 3805 | |||
XM_017010492.1 | XP_016865981.1 | 14 | 3849 | |||
GenBank mRNA | AB023153.1 | - | 14 | 3545 | ||
BC136420.1 | - | 13 | 3319 | 9020408 | ||
BC136421.1 | - | 13 | 3319 | 9020409 | ||
BC152464.1 | - | 14 | 3545 | 8862600 | ||
BX647493.1 | - | 2 | 1901 |