Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubash3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176860.5 | NP_789830.1 | 6 | 823 | ||
GenBank mRNA | AB075602.1 | - | 6 | 818 | ||
AK013361.1 | - | 6 | 891 | |||
AK034450.1 | - | 6 | 1014 | |||
AK035764.1 | - | 6 | 824 | |||
AK133948.1 | - | 6 | 821 | |||
AK150909.1 | - | 6 | 1390 | |||
AK151721.1 | - | 6 | 1391 | |||
AK151895.1 | - | 6 | 1024 | |||
AK152013.1 | - | 6 | 1024 | |||
AK154576.1 | - | 6 | 1045 | |||
BC053436.1 | - | 6 | 795 | 6510702 |