Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206927.1 | NP_001193856.1 | 6 | 1008 | ||
XM_011514318.2 | XP_011512620.1 | 6 | 1019 | |||
XM_011514319.2 | XP_011512621.1 | 6 | 1019 | |||
XM_011514320.2 | XP_011512622.1 | 4 | 782 | |||
XM_011514321.1 | XP_011512623.1 | 3 | 366 | |||
XM_017010325.1 | XP_016865814.1 | 6 | 1019 | |||
XM_017010326.1 | XP_016865815.1 | 6 | 1019 | |||
XM_017010327.1 | XP_016865816.1 | 6 | 1020 | |||
GenBank mRNA | AF356519.1 | - | 4 | 502 |