Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ptpn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177081.3 | NP_796055.1 | 3, 4 | 423 | ||
GenBank mRNA | AK041415.1 | - | 1, 2 | 1316 | ||
AK049175.1 | - | 2, 3 | 274 | |||
AK083305.1 | - | 3, 4 | 423 | |||
BC098475.1 | - | 3, 4 | 477 | 30044369 | ||
BC138194.1 | - | 2, 3 | 359 | 8861213 | ||
BC145190.1 | - | 2, 3 | 359 | 9053741 |