Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277239.1 | NP_001264168.1 | 5 | 499 | ||
NM_177086.4 | NP_796060.1 | 4 | 458 | |||
GenBank mRNA | AK036254.1 | - | 4 | 457 | ||
AK134793.1 | - | 4 | 457 | |||
AK140391.1 | - | 459 | ||||
AK140413.1 | - | 4 | 459 | |||
BC065110.1 | - | 4 | 446 | 30536600 | ||
BC079657.1 | - | 5 | 486 | 6853622 |