Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Glt1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177005.4 | NP_795979.2 | 8, 9 | 645 | ||
GenBank mRNA | AK050379.1 | - | 9, 10 | 765 | ||
AK077577.1 | - | 8, 9 | 647 | |||
BC115464.1 | - | 8, 9 | 644 |