Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029471.1 | - | 2 | 558 | ||
AK077292.1 | - | 2 | 505 | |||
AK160463.1 | - | 3 | 706 | |||
BB614316.1 | - | 2 | 558 |