Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025611.5 | NP_079887.3 | 2 | 870 | ||
GenBank mRNA | AK082134.1 | - | 1 | 1561 | ||
AK144856.1 | - | 1 | 1349 | |||
AK147156.1 | - | 2 | 628 | |||
AK147905.1 | - | 2 | 584 | |||
AK155454.1 | - | 1 | 1357 | |||
AK155583.1 | - | 2 | 874 | |||
AK166135.1 | - | 2 | 646 | |||
AK170580.1 | - | 2 | 873 | |||
AK170808.1 | - | 2 | 644 | |||
BC065096.1 | - | 1 | 506 | |||
BC138998.1 | - | 2 | 808 | 8862020 | ||
BC139000.1 | - | 2 | 808 | 8862022 |