Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpnpep3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177310.2 | NP_796284.1 | 1, 2 | 108 | ||
GenBank mRNA | AK042110.1 | - | 1, 2 | 147 | ||
AK088324.1 | - | 1, 2 | 108 | |||
AK158394.1 | - | 1, 2 | 127 | |||
BC137569.1 | - | 1, 2 | 92 | 8860588 | ||
BC144717.1 | - | 1, 2 | 93 | 9053265 |