Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177311.4 | NP_796285.2 | 4, 5 | 381 | ||
GenBank mRNA | AK019677.1 | - | 4, 5 | 370 | ||
BY715309.1 | - | 4, 5 | 367 |