Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtap7d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177293.3 | NP_796267.2 | 4 | 357 | ||
GenBank mRNA | AK030211.1 | - | 3 | 274 | ||
AK049164.1 | - | 4 | 347 | |||
BB610806.1 | - | 4 | 358 | |||
BC096764.1 | - | 4 | 358 | |||
BC118963.2 | - | 3 | 274 | 40095274 |