Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001372.3 | NP_001363.2 | 57, 58 | 11188 | ||
NM_004662.2 | NP_004653.2 | 3, 4 | 585 | |||
XM_011523703.1 | XP_011522005.1 | 57, 58 | 11152 | |||
XM_017024292.1 | XP_016879781.1 | 56, 57 | 11035 | |||
XM_017024293.1 | XP_016879782.1 | 46, 47 | 9096 | |||
GenBank mRNA | AB002355.2 | - | 38, 39 | 6866 | ||
AB290158.1 | - | 45, 46 | 9005 | |||
AF257737.1 | - | 57, 58 | 11178 | |||
AJ404468.1 | - | 57, 58 | 11188 | |||
AK302009.1 | - | 2, 3 | 401 | |||
BC128261.1 | - | 1, 2 | 125 | |||
BC128421.1 | - | 1, 2 | 125 | |||
BC153883.1 | - | 38, 39 | 6866 | |||
X99947.1 | - | 3, 4 | 585 |