Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3pxd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177364.3 | NP_796338.2 | 3 | 303 | ||
GenBank mRNA | AB430861.1 | - | 3 | 225 | ||
AK042261.1 | - | 3 | 289 | |||
AK043324.1 | - | 3 | 275 | |||
AK089330.1 | - | 3 | 275 | |||
AK090119.1 | - | 3 | 296 | |||
AK154898.1 | - | 3 | 261 | |||
AK163471.1 | - | 3 | 275 | |||
AK169978.1 | - | 3 | 295 | |||
AK171384.1 | - | 3 | 275 | |||
AK220175.1 | - | 3 | 292 | |||
BC020358.1 | - | 1 | 2370 | |||
BC115711.1 | - | 3 | 272 | |||
BC115764.1 | - | 3 | 272 | |||
BC141306.1 | - | 3 | 275 | 9055918 |