Gene Symbol: Slc9a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177353.3 | NP_796327.1 | 2 | 557 | ||
GenBank mRNA | AK041169.1 | - | 2 | 557 | ||
AK043187.1 | - | 2 | 579 | |||
AK078577.1 | - | 2 | 602 | |||
BC058750.1 | - | 2 | 563 | 6390297 |