Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zfp945 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110254.1 | NP_001103724.1 | 5 | 3486 | ||
NM_177358.4 | NP_796332.2 | 6 | 3607 | |||
GenBank mRNA | AK034838.1 | - | 1 | 1998 | ||
AK038672.1 | - | 1 | 790 | |||
AK041739.1 | - | 1 | 636 | |||
AK044245.1 | - | 1 | 405 | |||
AK164594.1 | - | 1 | 213 | |||
BC078460.1 | - | 6 | 3577 | 6851563 |