Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELAVL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001419.2 | NP_001410.2 | 3 | 391 | ||
GenBank mRNA | AK301013.1 | - | 3 | 393 | ||
BC003376.2 | - | 3 | 354 | 2901220 | ||
BT009793.1 | - | 2 | 224 | |||
BU542524.1 | - | 3 | 392 | |||
U38175.1 | - | 3 | 342 |