Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam167a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177628.4 | NP_808296.2 | 3 | 1070 | ||
GenBank mRNA | AK079469.1 | - | 1 | 146 | ||
BC060626.1 | - | 1 | 183 | |||
BC065085.1 | - | 3 | 853 | 6849398 | ||
BC094561.1 | - | 3 | 980 |