Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037719.2 | NP_001032808.2 | 6 | 745 | ||
NM_177584.3 | NP_808252.1 | 6 | 742 | |||
GenBank mRNA | AK041334.1 | - | 6 | 744 | ||
AY293285.1 | - | 6 | 729 | |||
BC108963.1 | - | 6 | 744 | 40046744 | ||
BC108964.1 | - | 6 | 747 | 40046745 |