Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pstk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039534.1 | NP_001034623.1 | 4 | 804 | ||
GenBank mRNA | AI173292.1 | - | 2 | 300 | ||
AK077585.1 | - | 4 | 805 | |||
BC030313.1 | - | 2 | 1296 | |||
BC110381.1 | - | 4 | 780 | 5149569 | ||
BC115503.2 | - | 4 | 805 |