Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gapt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177713.3 | NP_808381.1 | 3 | 657 | ||
GenBank mRNA | AK036534.1 | - | 3 | 658 | ||
AK155882.1 | - | 3 | 665 | |||
BC116629.1 | - | 1 | 504 | 40057168 | ||
BC118005.1 | - | 1 | 504 | 40057167 |