Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcmdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177722.3 | NP_808390.2 | 5 | 541 | ||
GenBank mRNA | AK031395.1 | - | 5 | 499 | ||
CA978963.1 | - | 3 | 296 |