Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dock11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009947.3 | NP_001009947.2 | 17 | 1970 | ||
GenBank mRNA | AB116935.2 | - | 17 | 1970 | ||
AK033257.1 | - | 5 | 1642 |