Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177765.3 | NP_808433.2 | 4, 5 | 717 | ||
GenBank mRNA | AK029565.1 | - | 4, 5 | 716 | ||
AK161519.1 | - | 4, 5 | 539 | |||
AM690757.1 | - | 3, 4 | 418 | |||
BC024578.1 | - | 3, 4 | 719 | |||
BC145749.1 | - | 3, 4 | 325 |