Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001334.2 | NP_001325.1 | 8 | 1143 | ||
GenBank mRNA | AK300112.1 | - | 6 | 624 | ||
AK308323.1 | - | 8 | 1148 | |||
BC049206.1 | - | 8 | 1096 | 5189657 | ||
X77383.1 | - | 8 | 1143 |