Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp874b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076791.2 | NP_001070259.1 | 4 | 350 | ||
GenBank mRNA | AK079330.1 | - | 2 | 56 | ||
AK137581.1 | - | 4 | 328 | |||
BY228006.1 | - | 2 | 133 |
Gene Symbol: Zfp874a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177712.4 | NP_808380.1 | 4 | 353 | ||
GenBank mRNA | AK049183.1 | - | 4 | 341 | ||
AK145787.1 | - | 4 | 353 | |||
BC080824.1 | - | 432 | 30458333 |