Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289747.1 | NP_001276676.1 | 14 | 2234 | ||
NM_001289748.1 | NP_001276677.1 | 13 | 2071 | |||
NM_001289749.1 | NP_001276678.1 | 11 | 1728 | |||
NM_177726.4 | NP_808394.1 | 13 | 2090 | |||
GenBank mRNA | AK037229.1 | - | 13 | 2080 | ||
AY159126.1 | - | 13 | 2080 | |||
AY177606.1 | - | 11 | 1718 | |||
AY177607.1 | - | 13 | 2071 | |||
BC112420.1 | - | 13 | 2067 | 40061385 | ||
BC113796.1 | - | 14 | 2211 | 40061392 |