Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DECR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001359.1 | NP_001350.1 | 5 | 588 | ||
XM_005250808.4 | XP_005250865.1 | 4 | 276 | |||
XM_011516880.2 | XP_011515182.1 | 6 | 583 | |||
XM_017013147.1 | XP_016868636.1 | 7 | 1243 | |||
XM_017013148.1 | XP_016868637.1 | 7 | 983 | |||
GenBank mRNA | AK300069.1 | - | 7 | 1103 | ||
BC105080.1 | - | 5 | 535 | 8144083 | ||
BC105082.1 | - | 5 | 535 | 8144085 | ||
BC143669.1 | - | 4 | 332 | |||
L26050.1 | - | 5 | 457 | |||
U49352.1 | - | 5 | 486 |