Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177879.5 | NP_808547.3 | 4 | 865 | ||
GenBank mRNA | AK030635.1 | - | 4 | 376 | ||
AK147578.1 | - | 4 | 910 | |||
BC060237.1 | - | 4 | 449 |