Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Catsper4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130030.1 | NP_001123502.1 | 3 | 213 | ||
NM_177866.4 | NP_808534.1 | 3 | 213 | |||
GenBank mRNA | AK077145.1 | - | 3 | 200 | ||
BC051630.1 | - | 3 | 201 | |||
BC108974.2 | - | 3 | 193 | 40047424 | ||
BC108975.1 | - | 3 | 193 | 40047425 | ||
BY104668.1 | - | 3 | 214 | |||
EU781510.1 | - | 3 | 188 | |||
EU781511.1 | - | 3 | 188 |