Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm5592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033782.3 | NP_001028954.1 | 2 | 436 | ||
GenBank mRNA | AK133295.1 | - | 2 | 437 | ||
BC147594.1 | - | 2 | 321 | 9056584 |
Gene Symbol: Gm4884 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC119562.1 | - | 2 | 392 | 40057558 |
Gene Symbol: Gm5114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177890.3 | NP_808558.2 | 2 | 438 | ||
GenBank mRNA | AK029498.1 | - | 2 | 439 |