Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EPB41L2 SIRNA Assays ›
Gene Symbol:
EPB41L2
Gene Name:
erythrocyte membrane protein band 4.1 like 2
Gene Aliases:
4.1-G, 4.1G
Chromosome Location:
Chr.6: 130829111 - 131063322 on Build GRCh38
UniGene ID:
Hs.486470
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
9951
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EPB41L2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135554.1
NP_001129026.1
3
718
NM_001135555.3
NP_001129027.1
3
731
NM_001199388.2
NP_001186317.1
3
731
NM_001252660.1
NP_001239589.1
3
698
NM_001431.3
NP_001422.1
3
718
XM_006715362.2
XP_006715425.1
3
749
XM_011535524.1
XP_011533826.1
3
718
XM_011535525.1
XP_011533827.1
3
718
XM_011535527.1
XP_011533829.1
3
718
XM_011535528.1
XP_011533830.1
3
718
XM_011535529.1
XP_011533831.1
3
718
XM_011535530.1
XP_011533832.1
3
718
XM_011535534.1
XP_011533836.1
3
718
XM_011535536.1
XP_011533838.1
3
718
XM_017010349.1
XP_016865838.1
3
746
XM_017010350.1
XP_016865839.1
3
680
XM_017010351.1
XP_016865840.1
3
742
XM_017010352.1
XP_016865841.1
3
584
XM_017010353.1
XP_016865842.1
3
718
XM_017010354.1
XP_016865843.1
3
718
XM_017010355.1
XP_016865844.1
3
746
XM_017010356.1
XP_016865845.1
3
718
XM_017010357.1
XP_016865846.1
3
680
XM_017010358.1
XP_016865847.1
3
718
XM_017010359.1
XP_016865848.1
3
746
XM_017010360.1
XP_016865849.1
3
584
XM_017010361.1
XP_016865850.1
3
746
XM_017010362.1
XP_016865851.1
3
680
XM_017010363.1
XP_016865852.1
3
708
GenBank mRNA
AB209522.1
-
3
629
AF027299.1
-
3
580
AK295124.1
-
3
660
AK307764.1
-
3
751
AL698619.1
-
3
642
AY047584.1
-
2
544
CR749262.1
-
3
660
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cortical actin cytoskeleton organization
actin binding
structural molecule activity
spectrin binding
PH domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-n6vvh:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0