Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgxb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178017.1 | NP_821136.1 | 11 | 2615 | ||
GenBank mRNA | AK088758.1 | - | 7 | 1443 | ||
BC048188.1 | - | 12 | 2729 | |||
BC049800.1 | - | 11 | 2615 | 6509446 |