Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps26b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178027.4 | NP_821170.1 | 1 | 583 | ||
GenBank mRNA | AK010160.1 | - | 1 | 519 | ||
AK031589.1 | - | 1 | 586 | |||
AK035321.1 | - | 1 | 536 | |||
AK150929.1 | - | 1 | 504 | |||
AK155578.1 | - | 1 | 544 | |||
AK165539.1 | - | 1 | 522 | |||
AK171154.1 | - | 1 | 542 | |||
AK185841.1 | - | 1 | 37 | |||
AK193143.1 | - | 1 | 37 | |||
AK194762.1 | - | 1 | 37 | |||
AK214365.1 | - | 1 | 33 | |||
BC049180.1 | - | 1 | 546 | 6314347 | ||
BC052721.1 | - | 1 | 507 | 6809853 | ||
BC062972.1 | - | 1 | 507 | 6809878 |