Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178110.2 | NP_835211.1 | 5 | 1164 | ||
GenBank mRNA | BC049095.1 | - | 5 | 1164 | 6416690 |