Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178387.3 | NP_848474.2 | 8 | 1260 | ||
GenBank mRNA | AK006916.1 | - | 7 | 938 | ||
BC050799.1 | - | 7 | 1162 | |||
BC120503.1 | - | 5 | 712 | |||
BC120505.1 | - | 5 | 712 | |||
BC144721.1 | - | 5 | 712 | |||
BC144723.1 | - | 5 | 712 |